Skip to Content
Merck

T110701

1,2-DIETHYL 3-METHYL PYRROLO(2,1-A)ISOQUINOLINE-1,2,3-TRICARBOXYLATE

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$448.92

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901


Select a Size

Change View
100 μG
$448.92

About This Item

Linear Formula:
C20H19NO6
CAS Number:
Molecular Weight:
369.378
MDL number:

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5200988SAB5200888SAB5200904
conjugate

unconjugated

conjugate

Allophycocyanin conjugate

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse, rat

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

clone

S327A-38, monoclonal

clone

S327A-38, monoclonal

clone

S327-95, monoclonal

clone

S327-95, monoclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service