Key Documents
SYNA341922
Synthware twin hose connecting adapter
joint: ST/NS 19/22
Synonym(s):
Synthware A341922, Adapter, twin connecting hose
Select a Size
1 EA
€24.80
€24.80
Check Cart for Availability
Select a Size
Change View
1 EA
€24.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€24.80
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A341922
Related Categories
General description
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)