Key Documents
SYNA342942
Synthware twin hose connecting adapter
joint: ST/NS 29/42
Synonym(s):
Synthware A342942, Adapter, twin connecting hose
Select a Size
1 EA
€28.80
€28.80
Check Cart for Availability
Select a Size
Change View
1 EA
€28.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€28.80
Check Cart for Availability
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A342942
Related Categories
General description
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)