Information on lapping is not provided by the vendor. The cone size is 40/38.
SLW5809/KIT
Pyrex® Vacuum filtration system for 47 mm membranes
complete assembly
Synonym(s):
Pyrex, SciLabware 5809/KIT
Select a Size
€499.00
Select a Size
About This Item
€499.00
Recommended Products
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
5809/KIT
SciLabware
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® Vacuum filtration system is a complete assembly of 300mL capacity funnel, support base, 1000mL conical flask, and metal retaining clamp.
- Manufactured using borosilicate glass
- Designed for use with membranes measuring 47mm in diameter.
- The system features a grade 3 porosity borosilicate glass sintered disc to support the membrane.
- For microbiology applications and HPLC solvents preparation.
- Components can also be ordered separately.
Features and Benefits
- Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
- For use with 47mm diameter membranes
- Complete assembly comprises of four components: - 5809/2 Graduated 300 ml capacity funnel - 5810/3 Support base with porosity 3 sintered filter disc and vacuum connection with 40/38 ground socket joint - 5810/4 1000 ml conical flask suitable for use under vacuum with 40/38 ground cone joint - 5810/5 Metal anodised retaining clamp
- Fully autoclavableIdeal for microbiology and preparation of HPLC solvents
- Support base has plastic screw thread tubing connector to accept 9mm bore vacuum tubing
- Each component is also available to order separately
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Quelle est la grandeur du rodage, est ce que le cône est du 40/38 ? Merci pour votre réponse
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service