Skip to Content
Merck
All Photos(2)

Key Documents

SLW3766/03

Pyrex® Filter funnel, Hirsch style with sintered glass disc

bowl size 14/23, porosity grade 3

Synonym(s):

Pyrex, SciLabware 3766/03

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€68.80

€68.80


Check Cart for Availability


Select a Size

Change View
1 EA
€68.80

About This Item

UNSPSC Code:
41104900
NACRES:
NB.43

€68.80


Check Cart for Availability

material

borosilicate glass

manufacturer/tradename

3766/03
SciLabware

bowl size

14/23

pore size

grade 3 porosity

General description

Named after the German chemist Robert Hirsch, these filter funnels are used for the vacuum filtration of very small volumes of precipitates.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service