Skip to Content
Merck
All Photos(1)

Documents

SLW1518/10

Pyrex® Media-Lab Bottles, plastic coated, with-out cap and pouring ring, with printed trace code

capacity 1000 mL

Synonym(s):

Pyrex, SciLabware 1518/10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

borosilicate glass bottle
round bottle

Joint

Joints threaded neck (GL45)

description

plastic coated

feature

graduations
without cap
with printed trace code
without pouring ring

packaging

pack of 10 ea

manufacturer/tradename

SciLabware 1518/10

H

225 mm

body diam.

101 mm

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Borosilicate glass with clear PU plastic coating to provide added protection against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Coating withstands steam sterilisation up to 135 deg. C

Features and Benefits

• Bottles only. Supplied without cap or pouring ring.• Complies with ISO 4796.• Borosilicate glass with clear PA12 plastic coating to provide added protection against mechanical damage.• Reduces the hazard of flying glass and liquid splashes in the event of breakage.• Printed with Trace Code for downloadable lot certificate.• This product is suitable for steam sterilisation at 121 or 134 °C

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service