SLW1025/22SC
Pyrex® Beakers, jacketed with PTFE screwthread
1000 mL
Synonym(s):
Pyrex, SciLabware 1025/22SC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
1025/22SC
SciLabware
H
215 mm , internal
I.D.
80 mm
O.D.
110 mm
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Useful for maintaining samples at a constant temperature
- With integral jacket through which a temperature controlling medium can be circulated
- Manufactured from heat resistant Pyrex® borosilicate glass
- With PTFE screwthread connectors for simple and safe connection of 9mm bore tubing
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service