Skip to Content
Merck
All Photos(1)

Key Documents

QBD10932

Sigma-Aldrich

m-dPEG®48-MAL

>95% (HPLC)

Synonym(s):

m-PEG48-MAL, mPEG-maleimide, mPEG2000-maleimide, mPEG48-maleimide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€486.00

€486.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€486.00

About This Item

Empirical Formula (Hill Notation):
C104H202N2O51
Molecular Weight:
2296.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€486.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(C(=O)C=CC1=O)CCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC

InChI key

DBVPVJXDAXLVIM-UHFFFAOYSA-N

Features and Benefits

m-dPEG48-MAL is a thiol-reactive, monodispersed PEGylation reagent used for surface and biomolecule modification. One end of the PEG molecule is methyl-terminated. The opposite end possesses a 3-maleimido propanoic acid functional group that connects to the PEG spacer via an amide bond. The maleimide moiety reacts with sulfhydryl groups by the thiol-Michael addition reaction. The single molecular weight, discrete-length PEG (dPEG) chain is 152 atoms (178.6 Å) long, including the reactive site on the maleimide ring. The dPEG48 is a monodisperse equivalent of the polymeric PEG2000.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas M Molino et al.
Biomacromolecules, 13(4), 974-981 (2012-03-16)
Self-assembling protein nanocapsules can be engineered for various bionanotechnology applications. Using the dodecahedral scaffold of the E2 subunit from pyruvate dehydrogenase, we introduced non-native surface cysteines for site-directed functionalization. The modified nanoparticle's structural, assembly, and thermostability properties were comparable to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service