Skip to Content
Merck
All Photos(1)

Key Documents

880138P

Avanti

DSPE-PEG(2000) Carboxy NHS

Avanti Research - A Croda Brand

Synonym(s):

DSPE-PEG(2000) Carboxy NHS, 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[carboxy(polyethylene glycol)-2000, NHS ester] (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€372.00

€372.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€372.00

About This Item

Empirical Formula (Hill Notation):
C135H260N2NaO57P
CAS Number:
Molecular Weight:
2877.45
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€372.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

package of 1 × 10 mg

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

Application


  • Enhanced detection of myeloma cells: DSPE-PEG(2000) Carboxy was used to create fluorescence labeled XT5 modified nano-capsules, enhancing the sensitivity of detecting myeloma cells. This application demonstrates its potential in biomedical diagnostics and targeted therapy (Norouz Dizaji et al., 2022).

  • Improving drug encapsulation efficiency: The use of DSPE-PEG(2000) Carboxy in pre-formed liposomes for drug delivery showcases a method that significantly improves carboplatin encapsulation, highlighting its role in optimizing chemotherapeutic drug formulations (Wehbe et al., 2017).

  • Targeted drug delivery for cardiovascular diseases: DSPE-PEG(2000) Carboxy was integral in the development of PEGylated-nanoliposomal formulations of clusterin, aimed at treating amyloidogenic light chain-induced endothelial dysfunction, underscoring its utility in targeted cardiovascular therapies (Guzman-Villanueva et al., 2018).

Packaging

5 mL Amber Glass Screw Cap Vial (880138P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service