Skip to Content
Merck
All Photos(1)

Documents

QBD10631

Sigma-Aldrich

Bis-MAL-Lysine-dPEG®4-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H45F4N5O13
Molecular Weight:
843.77
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C(NC(CCN1C(C=CC1=O)=O)=O)CCCCNC(CCN2C(C=CC2=O)=O)=O)NCCOCCOCCOCCOCCC(OC3=C(C(F)=CC(F)=C3F)F)=O

Features and Benefits

Bis-MAL-Lysine-dPEG®4-TFP ester is a unique crosslinker containing two maleimide groups, one on each amine of lysine, a 4-unit, single molecular weight dPEG® spacer arm, and a 2,3,5,6-tetrafluorophenyl (TFP) ester. Each maleimide group reacts with a sulfhydryl group, and the TFP ester reacts with a free amine. The single molecular weight dPEG® spacer is hydrophilic and non-immunogenic. This reagent can be used to modify or form conjugates with antibodies, antibody fragments, peptides, and small molecules.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service