Skip to Content
Merck
All Photos(1)

Key Documents

QBD10195

Sigma-Aldrich

Biotin-dPEG®11-MAL

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€359.00

€359.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€359.00

About This Item

Empirical Formula (Hill Notation):
C41H71N5O16S
Molecular Weight:
922.09
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

€359.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: Biotinylations
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCN1C(C=CC1=O)=O)=O)CCCCC2C3NC(NC3CS2)=O

InChI key

FMIDVLYXFRCQAD-UHFFFAOYSA-N

Features and Benefits

Biotin-dPEG®11-MAL takes advantage of the well-known, specific Michael reaction between a maleimide group and a free sulfhydryl to allow site-specific biotin labeling of molecules in aqueous media within the pH range of 6.5 – 7.5. The single molecular weight dPEG® spacer (50.5 Å) provide precise spacing between the biotin moiety and the labeled molecule. The dPEG® spacer imparts water solubility to the labeled molecule while reducing immunogenicity, aggregation, and precipitation of labeled biomolecules or other labeled compounds.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service