Skip to Content
Merck
All Photos(1)

Key Documents

OCNQCK645

Sigma-Aldrich

EDC Conjugation Kit for Carboxylic Cd-based Core/Shell Quantum Dots

fluorescence λem 645 nm, for 4 conjugation

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€480.00

€480.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€480.00

About This Item

UNSPSC Code:
43211915
NACRES:
NA.23

€480.00


Estimated to ship on01 May 2025


Request a Bulk Order

fluorescence

λem 645 nm

Looking for similar products? Visit Product Comparison Guide

General description

Components:
  • QSH
  • EDC
  • Coupling Buffer
  • Quenching Buffer
  • Storage Buffer
  • Tubes 4 x 1.7 mL

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service