OCNQAK665
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots
fluorescence λem 665 nm, for 4 conjugation
About This Item
Recommended Products
fluorescence
λem 665 nm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- QSA
- Sulfo-SMCC
- Traut′s Reagent
- Coupling Buffer
- Storage Buffer
- Quenching Buffer
- Tubes 4 x 1.7 mL
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service