Skip to Content
Merck
All Photos(1)

Key Documents

OCNIAK30

Sigma-Aldrich

SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles

30 nm, amine functionalized, 5 mg/mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€356.00

€356.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€356.00

About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

€356.00


Estimated to ship on26 March 2025


Request a Bulk Order

form

nanoparticles

concentration

5 mg/mL

particle size

30 nm

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide nanoparticles are magnetic single crystals and can be used as magnetic resonance imaging contrast reagents for in-vivo imaging , drug delivery vehicle, hyperthermia genertor for therapy, adjuvant for vaccine, and magnetic sensing probe for in-vitro diagnostics. These amine terminated iron oxide nanocrystals of various diameters (10 nm-30 nm) are now available in an easy-to-use kit format to enable researchers to conjugate proteins/ligands of their own choice to these iron oxide nanoparticles. The kit is used to specifically conjugate iron oxide nanoparticles and thiol containing ligands with low non-specific binding. It contains sufficient reagents & components for performing 5 conjugation reactions using 1 mg iron oxide nanoparticles per reaction. The iron oxide nanoparticles are activated using SMCC (succinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate) followed by conjugation to thiol groups that are present on the target protein/ligands such as bovine serum albumin, streptavidin and immunoglobulin.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service