Skip to Content
Merck
All Photos(1)

Key Documents

NCXAUSH50

Sigma-Aldrich

Gold Nanospheres

Silica-Shelled, 50 nm, NanoXact, 1 mg/mL in ethanol, Silica (Silanol)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€292.00
10 ML
€1,940.00

€292.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€292.00
10 ML
€1,940.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

€292.00


Check Cart for Availability

Request a Bulk Order

form

nanospheres

concentration

1 mg/mL in ethanol

particle size

50 nm

Looking for similar products? Visit Product Comparison Guide

Application

Gold Nanospheres (silica-shelled, 50 nm, NanoXact, 1 mg/mL in ethanol, Silica (Silanol)) can be used for a variety of potential applications such as photothermal therapy, surface enhanced Raman spectroscopy, and biosensors.[1][2][3]
Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our silica shelled gold nanoparticles possess robust surface and exhibit high stability in variety of solvents. The silica shell offers a versatile surface for attaching functional groups or creating hydrophobic & hydrophilic surfaces. The presence of silica shell on the surface of gold nanoparticles also allows their easier deposition onto substrates and incorporation into different solvents, resins, and curable materials while protecting them from aggregation and preserving their optical properties.

Storage and Stability

DO NOT FREEZE. Store at 2-8C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile one-pot synthesis of gold nanoparticles stabilized with bifunctional amino/siloxy ligands
Zhu H, et al.
Journal of Colloid and Interface Science, 287(1), 360-365 (2005)
The potential use of the enhanced nonlinear properties of gold nanospheres in photothermal cancer therapy
Huang X, et al.
Lasers in Surgery and Medicine, 39(9), 747-753 (2007)
Exceptionally high payload of doxorubicin in hollow gold nanospheres for near-infrared light-triggered drug release
You J, et al.
ACS Nano, 4(2), 1033-1041 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service