Skip to Content
Merck
All Photos(1)

Documents

MT51302069

Mettler-Toledo technical buffer

pH 4.01, volume 30 x 20 mL , sachets

Synonym(s):

liquid pH 4.01 buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191600
NACRES:
NA.25

sterility

non-sterile

form

liquid

manufacturer/tradename

Mettler-Toledo 51302069

volume

30 x 20 mL , sachets

pH

4.01

Looking for similar products? Visit Product Comparison Guide

General description

METTLER TOLEDO certified pH buffer solutions are traceable to NIST primary standards and come with a quality inspection certificate, which guarantees the stated values and traceability. We offer convenient package sizes from single-use 20 mL pouches to 20 L Cubitainers. Our variety packs of pH 4, 7 and 10 buffers are offered in both 20 mL pouches and convenient 500 mL bottles.

Features and Benefits

  • High-quality Mettler-Toledo Rainbow I buffers for accurate sensor calibration and maintenance.
  • Small volume packaging (single-use sachets) ensures guaranteed fresh solutions and ease of use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service