Skip to Content
Merck
All Photos(2)

Key Documents

I26809

Sigma-Aldrich

Isopropyl sulfide

99%

Synonym(s):

Diisopropyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€241.00
100 G
€486.00

€241.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
€241.00
100 G
€486.00

About This Item

Linear Formula:
[(CH3)2CH]2S
CAS Number:
Molecular Weight:
118.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€241.00


Only 0 left in stock

Request a Bulk Order

Assay

99%

refractive index

n20/D 1.4379 (lit.)

bp

120 °C/763 mmHg (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CC(C)SC(C)C

InChI

1S/C6H14S/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

XYWDPYKBIRQXQS-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanisław Popiel et al.
Analytica chimica acta, 837, 52-63 (2014-07-09)
Novel solid-phase microextraction (SPME) fibres containing methyl, ethyl, butyl acrylate and methacrylate were first prepared by a sol-gel technique and investigated for determination of selected organoarsenic compounds (lewisite, methyldichloroarsine, phenyldichloroarsine, diphenylchloroarsine and triphenylarsine) from water samples. The influence of sorption

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service