This is a product of Goodfellow. The length and diameter of the carbon fiber used in the preparation of this product has not been deternined.
GF11766540
Polyamide - Nylon 6, 6 - 30% Carbon Fiber Reinforced
(PA 6, 6 - 30% CFR), granule, 3 mm nominal granule size, weight 500 g, black
Synonym(s):
AM396310
Select a Size
€303.00
Select a Size
About This Item
€303.00
Recommended Products
form
granular
manufacturer/tradename
Goodfellow 117-665-40
weight
500 g
color
black
nominal granule size
3 mm
Related Categories
General description
Legal Information
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the length and diameter of carbon fiber in the pellets
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service