Skip to Content
Merck
All Photos(1)

Key Documents

E1055

Sigma-Aldrich

Epichlorohydrin

99%

Synonym(s):

(±)-Epichlorohydrin, (±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein:
79785
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.2 (vs air)

vapor pressure

13.8 mmHg ( 21.1 °C)

Assay

99%

autoignition temp.

779 °F

expl. lim.

21 %

refractive index

n20/D 1.438 (lit.)

bp

115-117 °C (lit.)

mp

−57 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

ClCC1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

BRLQWZUYTZBJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karnkamol Trisopon et al.
Pharmaceutics, 12(6) (2020-06-11)
A new co-processed, rice starch-based excipient (CS) was developed via a spray-drying technique. Native rice starch (RS) was suspended in aqueous solutions of 10%-15% cross-linked carboxymethyl rice starch (CCMS) and 0.5%-6.75% silicon dioxide (in the form of sodium silicate), before
A K Giri
Mutation research, 386(1), 25-38 (1997-03-01)
Epichlorohydrin (ECH) is one of the more commercially important aliphatic epoxides used extensively as an industrial intermediate, a laboratory reagent, and as an insecticide. It is a volatile, colourless liquid with an ethereal odour. It behaves as an alkylating agent.
An evaluation of the genetic toxicity of epichlorohydrin. A report of an expert group of the International Commission for Protection against Environmental Mutagens and Carcinogens.
R J Srám et al.
Mutation research, 87(3), 299-319 (1981-11-01)
Swetha Inturi et al.
ACS nano, 9(11), 10758-10768 (2015-10-22)
Notwithstanding rapid advances of nanotechnology in diagnostic imaging and drug delivery, the engineered nanocarriers still exhibit substantial lack of hemocompatibility. Thus, when injected systemically, nanoparticles are avidly recognized by blood leukocytes and platelets, but the mechanisms of immune recognition are
Jingyun Wang et al.
Biomacromolecules, 13(2), 559-564 (2012-01-20)
The stability of a paper-immobilized antibody was investigated over a range of temperatures (40-140 °C) and relative humidities (RH, 30-90%) using both unmodified filter paper and the same paper impregnated with polyamide-epichlorohydrin (PAE) as supports. Antibody stability decreased with increasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service