Skip to Content
Merck
All Photos(2)

Key Documents

DWKWGW0255V

Azlon Gram Stain Wash Bottles with Driplok vapor venting

low-density polyethylene bottle, capacity 250 mL

Synonym(s):

WGW0255V

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43
Pricing and availability is not currently available.

material

low-density polyethylene bottle

packaging

pkg of 1 ea

manufacturer/tradename

Azlon WGW0255V

capacity

250 mL

Application

Ideal for use with Gram-stain solutions

Features and Benefits

Set of 5 wash bottles for use with Crystal Violet solution, Lugols iodine solution, Gram iodine solution, Gram colour/de-colourising solution, Safranine solution

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service