Skip to Content
Merck
All Photos(1)

Key Documents

1.00567

Sigma-Aldrich

Lugol′s solution

stabilized with PVP for the Gram staining

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€74.10
2.5 L
€150.00

€74.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€74.10
2.5 L
€150.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.74

€74.10


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

2.0 (20-22 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The Lugol′s solution stabilized with PVP, is a ready-to-use solution used in bacteriology, for example, for the staining of Gram bacteria. It contains a complex of Iodine-Polyvinylpyrrolidon (PVP) and no free iodine, which stabilizes the solution and reduces the evaporation of iodine. During Gram-staining, the iodine from the Lugol′s solution and crystal violet (Cat. no. 109218) form a stable complex that will not be released by decolorisation. Thus, Gram-positive bacteria will appear blue-violet when analyzed microscopically, wheras Gram-negative bacteria can be counterstained with a safranine solution (Product number 1.09217) and will appear pink to red.
1 L of the Lugol′s solution stabilized with PVP is capable of staining up to 500 slides. This product is registered as IVD and CE marked.For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
gram-positve microorganisms: blue-violet
gram-negative microorganisms: pink to red

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service