Key Documents
DWK73808-33430
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
33-430
Select a Size
1 PKG
€124.00
€124.00
Check Cart for Availability
Select a Size
Change View
1 PKG
€124.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
€124.00
Check Cart for Availability
material
PTFE/silicone liner
black polypropylene screw cap (33-430 thread)
packaging
pkg of 1 packs (48 caps)
manufacturer/tradename
Kimble® 73808-33430
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Closures are ideal for repeated autoclaving
  • Welded liner technology eliminates the possibility of glue contamination
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the heat capacity of these lids? Can they withstand autoclave temperatures/pressure?
    1 answer
    1. This cap is suitable for use up to 135°C and may be repeatedly autoclaved at 121°C at 15 psig for 20 minutes. To prevent damage to containers, caps should be loose during autoclaving to allow for venting.
      Helpful?