Key Documents
DWK73802-13415
KIMBLE® solid cap with PTFE faced rubber liner
black phenolic cap, cap for 13-415
Select a Size
1 EACH
€169.00
€169.00
Check Cart for Availability
Select a Size
Change View
1 EACH
€169.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
€169.00
Check Cart for Availability
material
PTFE/rubber liner
black phenolic cap
sterility
non-sterile
feature
class A
packaging
case of 500 ea
manufacturer/tradename
Kimble® 73802-13415
fitting
cap for 13-415
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavabl
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirement
Features and Benefits
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)