DWK218067358
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (5,000 mL), non-sterile
Synonym(s):
218067358
Select a Size
€280.00
Select a Size
About This Item
€280.00
Recommended Products
material
round amber glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 1 ea
manufacturer/tradename
DURAN® 218067358
bottle capacity
(5,000 mL)
General description
Features and Benefits
- Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
- The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
- Very durable and chemically resistant amber colour
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)