Skip to Content
Merck
All Photos(1)

Key Documents

DWK218015414

Duran® Laboratory Bottle

clear, graduated, GL45, 1000mL, PBT screw cap, ETFE pouring ring, 10EA

Synonym(s):

218015414

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
€244.00

€244.00


Check Cart for Availability


Select a Size

Change View
10 EACH
€244.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

€244.00


Check Cart for Availability

material

clear bottle

Joint

Joints threaded neck (GL45)

sterility

non-sterile

feature

graduated
with screw cap to drain contents WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 218015414

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN® borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN® Original GL 45 laboratory bottles are available complete with high temperature screw caps and pouring rings. The red polybutylene terephthalate (PBT) GL 45 threaded caps with PTFE faced silicone cap liner and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. All components are fully autoclavable at 121 or 134 °C and suitable for dry heat sterilization up to 180 °C. All components are fully compliant with current international food contact regulations such as (EU) No 10/2011.

Application

Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Liquid handling reservoir, Boro 3.3, PBT, ETFE, high temperature

Features and Benefits

  • Manufactured from DURAN™ borosilicate glass 3.3. DURAN™ is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • Supplied complete with red polybutylene terephthalate (PBT) GL 45 threaded cap with PTFE faced silicone cap liner and ETFE pouring ring for drip free pouring
  • All components are fully autoclavable at 121 or 134 °C
  • All components are suitable for dry heat sterilization up to 180 °C
  • Permanent Retrace Code on each bottle for full manufacturing lot traceability
  • The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel
  • Suitable for long-term storage

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service