Skip to Content
Merck
All Photos(1)

Key Documents

D56004

Sigma-Aldrich

3,5-Dichloroanthranilic acid

97%

Synonym(s):

2-Amino-3,5-dichlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2-2-(NH2)CO2H
CAS Number:
Molecular Weight:
206.03
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

gray to brown

mp

227-230 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1c(Cl)cc(Cl)cc1C(O)=O

InChI

1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,10H2,(H,11,12)

InChI key

KTHTXLUIEAIGCD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Kouzai et al.
The New phytologist, 217(2), 771-783 (2017-10-20)
Rhizoctonia solani is a soil-borne fungus causing sheath blight. In consistent with its necrotrophic life style, no rice cultivars fully resistant to R. solani are known, and agrochemical plant defense activators used for rice blast, which upregulate a phytohormonal salicylic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service