Skip to Content
Merck
All Photos(1)

Key Documents

D22908

Sigma-Aldrich

9,10-Diaminophenanthrene

97%

Synonym(s):

9,10-Phenanthrenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€240.00

€240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€240.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€240.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

164-166 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1c(N)c2ccccc2c3ccccc13

InChI

1S/C14H12N2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8H,15-16H2

InChI key

VPRFQZSTJXHBHL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberley J Davis et al.
Dalton transactions (Cambridge, England : 2003), 47(38), 13573-13591 (2018-09-13)
Two different series of nickel Schiff base complexes were prepared as part of a study aimed at discovering new compounds with high affinity and selectivity for quadruplex DNA (qDNA). The new complexes were prepared by modification of a literature method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service