Skip to Content
Merck
All Photos(3)

Key Documents

D144002

Sigma-Aldrich

3-(Dimethylamino)phenol

97%

Synonym(s):

N,N-Dimethyl-3-aminophenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€51.00
25 G
€111.00

€51.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
€51.00
25 G
€111.00

About This Item

Linear Formula:
(CH3)2NC6H4OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
774631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
crystals
Assay:
97%

€51.00


Estimated to ship on29 March 2025


Request a Bulk Order

Quality Level

Assay

97%

form

crystals

mp

82-84 °C (lit.)

SMILES string

CN(C)c1cccc(O)c1

InChI

1S/C8H11NO/c1-9(2)7-4-3-5-8(10)6-7/h3-6,10H,1-2H3

InChI key

MESJRHHDBDCQTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bozena Bukowska et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(8), 1574-1580 (2007-07-17)
3-(Dimethylamino)phenol (3-DMAP) exists in the environment as a transformation product of ureic herbicides and may also be considered as a derivative of phenoxyherbicides. In this study, the activity of glutathione peroxidase, catalase and superoxide dismutase, as well as the level
Anežka Tichá et al.
The Journal of biological chemistry, 292(7), 2703-2713 (2017-01-11)
Rhomboid proteases are increasingly being explored as potential drug targets, but their potent and specific inhibitors are not available, and strategies for inhibitor development are hampered by the lack of widely usable and easily modifiable
Avi Weissberg et al.
Journal of chromatography. A, 1512, 71-77 (2017-07-18)
A methodology for sensitive determination of sarin (GB), soman (GD) and cyclosarin (GF) chemical warfare agents in aqueous media was developed. The method incorporates direct derivatization with 2-[(dimethylamino)methyl]phenol (2-DMAMP), a commercially available, water-soluble reagent, followed by LC-ESI-MS/MS analysis in the
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service