Skip to Content
Merck
All Photos(3)

Key Documents

249394

Sigma-Aldrich

8-Hydroxyjulolidine

96%

Synonym(s):

2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€121.00
5 G
€403.00

€121.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€121.00
5 G
€403.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
Beilstein:
1530423
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€121.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

powder or crystals

composition

Carbon 72.7-79.6%, Nitrogen 7.1-7.7%

color

light brown

mp

132-134 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2CCCN3CCCc1c23

InChI

1S/C12H15NO/c14-11-6-5-9-3-1-7-13-8-2-4-10(11)12(9)13/h5-6,14H,1-4,7-8H2

InChI key

FOFUWJNBAQJABO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The carboxaldehyde derivative of 8-hydroxyjulolidine is useful as a chromophore in fluorescence chemosensor and chemosensors.[1]

Application

8-Hydroxyjulolidine has been used in the electrochemical switching fluorescence study.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new multifunctional Schiff base as a fluorescence sensor for Al 3+ and a colorimetric sensor for CN- in aqueous media: an application to bioimaging
Lee SA, et al.
Dalton Transactions, 43(18), 6650-6659 (2014)
Electrochemical switching fluorescence emission in rhodamine derivatives
Cizkova M, et al.
Electrochimica Acta, 260, 589-597 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service