Skip to Content
Merck
All Photos(1)

Key Documents

D131407

Sigma-Aldrich

2,3-Dimethoxybenzoic acid

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€41.50
25 G
€99.60

€41.50


Estimated to ship on12 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€41.50
25 G
€99.60

About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
2210858
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€41.50


Estimated to ship on12 May 2025


Request a Bulk Order

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

mp

120-122 °C (lit.)

SMILES string

COc1cccc(C(O)=O)c1OC

InChI

1S/C9H10O4/c1-12-7-5-3-4-6(9(10)11)8(7)13-2/h3-5H,1-2H3,(H,10,11)

InChI key

FODBVCSYJKNBLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service