CLS824050
Pyrex® pour-out centrifuge tube with round bottom
capacity 50 mL
Synonym(s):
Corning® 8240
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
sterility
non-sterile
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8240-50
O.D. × L
29 mm × 116 mm
capacity
50 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Pyrex® pour-out centrifuge tube is made of low-expansion borosilicate glass.
- Features a pour-out spout for easy decanting.
- Autoclavable - The product withstands repeated standard autoclave cycles of 121°C at 15 psi for 15 minutes.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service