Key Documents
Z564907
Buchi® diagonal condenser, for Rotavapor® evaporators
for R210/215, glass
Select a Size
1 EA
€822.00
€822.00
Check Cart for Availability
Select a Size
Change View
1 EA
€822.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€822.00
Check Cart for Availability
material
glass
manufacturer/tradename
Büchi 46988
General description
Condensers for R200/205 and R210/215 come with hose barbs and drain sleeve. Replacement condensers do not include stopcocks.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)