CLS70795N
Pyrex® serological pipettes, disposable, non-sterile, unplugged
volume 5.0 mL
Synonym(s):
pipet
Select a Size
€457.00
Select a Size
About This Item
€457.00
Recommended Products
material
borosilicate glass
sterility
non-sterile
feature
DNase free: no
RNase free: no
graduations
subdivision
packaging
pack of 240 ea
case of 960 ea
manufacturer/tradename
Corning 7079-5N
volume measuring range
5.0 mL
capacity
5.0 mL
tip size
, Narrow
Related Categories
Features and Benefits
- Disposable PYREX® glass pipettes are calibrated "to deliver" (TD) with blow-out
- Long, slender, tapered tips of pipettes make pipetting faster and easier
- All pipettes have negative graduations and fire polished tips for burr-free, even flow rates
- Nonsterile, unplugged and bulk packed.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)