CLS6949E
Corning® organic chemistry glassware kit
with 19/22 joints
Synonym(s):
Corning® 6949E and 6949K
About This Item
Recommended Products
manufacturer/tradename
Corning 6949E
Looking for similar products? Visit Product Comparison Guide
General description
Components:
1. Distilling column
2. West condenser
3. Separatory funnel, 125 mL
4. Bleed tube
5. Rubber thermometer holder
6. Thermometer adapter
7. Round-bottom flask, 100 mL
8. Round-bottom flask, 250 mL
9. 3-way adapter, 3 ground-glass joints
10. Round-bottom flask, 50 mL
11. Vacuum adapter
12. Claisen adapter
13. Round-bottom flask, 25 mL
14. PYREX stopper
15. Round-bottom flask, 500 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service