Key Documents
CLS53402L
Pyrex® heavy wall graduated filtering flask, with tubulation
12.5 mm O.D. hose-connection, capacity 2,000 mL
Synonym(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 5340
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper9
sterility
non-sterile
feature
closure type Rubber stopper
packaging
pack of 1
case of 12
manufacturer/tradename
Corning 5340-2L
volume measuring range
600-2,000 mL
capacity
2,000 mL
General description
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)