C83708
L-Citrulline
98%
Synonym(s):
(S)-2-Amino-5-ureidopentanoic acid, L-2-Amino-5-ureidovaleric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19
Beilstein:
1725416
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Recommended Products
Assay
98%
form
crystals
application(s)
peptide synthesis
SMILES string
N[C@@H](CCCNC(N)=O)C(O)=O
InChI
1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1
InChI key
RHGKLRLOHDJJDR-BYPYZUCNSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C C Reparon-Schuijt et al.
Arthritis and rheumatism, 44(1), 41-47 (2001-02-24)
To understand the regulation of anti-citrulline-containing peptide antibody (anti-CCP) production in rheumatoid arthritis (RA), production of anti-CCP by B cells derived from peripheral blood (PB), bone marrow (BM), and synovial fluid (SF) was examined. Purified PB and SF B cells
Koen M J Janssen et al.
Current opinion in rheumatology, 25(2), 241-247 (2013-02-02)
This article reviews the link between periodontitis and rheumatoid arthritis (RA) with regard to similarities in genetic risk factors and immunopathogenesis. Emphasis is paid to the potential role of the periodontal pathogen Porphyromonas gingivalis in the etiopathogenesis of both periodontitis
Lars Klareskog et al.
Advances in immunology, 118, 129-158 (2013-05-21)
Rheumatoid arthritis (RA) represents a disease where we have recently acquired new knowledge on etiology and molecular pathogenesis, by combining data from studies on genetic end environmental determinants of disease with molecular and cellular immunology. This combined approach has provided
P Crenn et al.
Gastroenterology, 119(6), 1496-1505 (2000-12-13)
No blood marker assessing the functional absorptive bowel length has been identified. Plasma citrulline, a nonprotein amino acid produced by intestinal mucosa, is one candidate. We tested this hypothesis in adult patients with the short-bowel syndrome, whose condition can lead
Cecilia Orellana et al.
Annals of the rheumatic diseases, 73(4), 752-755 (2013-07-28)
To study the impact of parity history on the risk of antibodies to citrullinated peptide antigens (ACPA) positive and ACPA-negative rheumatoid arthritis (RA), in different age-groups. Data from a population-based case-control study of female incident RA cases were analysed (2035
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service