Skip to Content
Merck
All Photos(1)

Key Documents

27505

Sigma-Aldrich

D-Citrulline

≥99.0%, for peptide synthesis

Synonym(s):

(R)-2-Amino-5-ureidopentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€198.00

€198.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€198.00

About This Item

Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19
Beilstein:
1725415
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€198.00


Check Cart for Availability

Request a Bulk Order

Product Name

D-Citrulline, ≥99.0%

Quality Level

Assay

≥99.0%

form

powder

optical activity

[α]20/D −20.5±2.0°, c = 2% in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

N[C@H](CCCNC(N)=O)C(O)=O

InChI

1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m1/s1

InChI key

RHGKLRLOHDJJDR-SCSAIBSYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvain V Costes et al.
PLoS computational biology, 3(8), e155-e155 (2007-08-07)
Several proteins involved in the response to DNA double strand breaks (DSB) form microscopically visible nuclear domains, or foci, after exposure to ionizing radiation. Radiation-induced foci (RIF) are believed to be located where DNA damage occurs. To test this assumption
Nikhil Vadhavkar et al.
Radiation research, 182(3), 273-281 (2014-07-31)
In contrast to the classic view of static DNA double-strand breaks (DSBs) being repaired at the site of damage, we hypothesize that DSBs move and merge with each other over large distances (μm). As X-ray dose increases, the probability of
S V Costes et al.
Mutation research, 704(1-3), 78-87 (2010-01-12)
DNA damage sensing proteins have been shown to localize to the sites of DNA double strand breaks (DSB) within seconds to minutes following ionizing radiation (IR) exposure, resulting in the formation of microscopically visible nuclear domains referred to as radiation-induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service