Skip to Content
Merck
All Photos(2)

Key Documents

C6257

Sigma-Aldrich

Creatinine hydrochloride

≥97%

Synonym(s):

2-Amino-1-methyl-2-imidazolin-4-one hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€95.30

€95.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€95.30

About This Item

Empirical Formula (Hill Notation):
C4H7N3O · HCl
CAS Number:
Molecular Weight:
149.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€95.30


Check Cart for Availability

Request a Bulk Order

Assay

≥97%

form

powder

SMILES string

Cl.CN1CC(=O)NC1=N

InChI

1S/C4H7N3O.ClH/c1-7-2-3(8)6-4(7)5;/h2H2,1H3,(H2,5,6,8);1H

InChI key

PJYONPGZTHDDJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Catalytic hydrogenation of acylguanidines
  • Strong binding of alkylguanidinium ions by molecular tweezers for a selective arginine receptor molecule
  • Investigations of drug distribution in the body after oral administration to rats

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elke Gasthuys et al.
Scientific reports, 9(1), 19699-19699 (2019-12-25)
Early diagnosis of kidney diseases in avian species is limited. Endogenous markers currently used in avian practice are not sensitive enough to identify early kidney failure. Consequently, alternative markers should be evaluated. To be able to evaluate these alternative markers
Bernhard Brauner et al.
International journal of pharmaceutics, 579, 119158-119158 (2020-02-23)
Recurring infections and increasing resistances continue to complicate treatment of urinary tract infections. To investigate alternative treatment options, trimethoprim loaded micro- (D[4;3] of 1-9 µm) and nanoparticles (Z-Avg of 200-400 nm) were prepared from two types of poly(d,l-lactic-co-glycolic acid) (PLGA) for instillative
Victor K Langsi et al.
Journal of chromatography. A, 1506, 37-44 (2017-05-27)
A rapid chromatographic method for the simultaneous determination of uric acid (UA) and creatinine (Cr) in human urine is described, using a non-bonded 1.7μm thin-shell (TS1.7-100nm) silica particle prepared by the seeded-growth approach. The new shell particle was characterised by
Fiorella Lucarini et al.
International journal of environmental research and public health, 17(13) (2020-07-09)
Restrictions on the use of bisphenol A (BPA) in consumer products led to its replacement by various bisphenol (BP) analogues, yet young children's exposure to these analogues has been poorly characterized so far. This study aimed to characterize infants' and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service