Skip to Content
Merck
All Photos(1)

Key Documents

532177

Sigma-Aldrich

3,4-Dichlorobenzonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€46.40

€46.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€46.40

About This Item

Linear Formula:
Cl2C6H3CN
CAS Number:
Molecular Weight:
172.01
Beilstein:
2326352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€46.40


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

74-78 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(cc1Cl)C#N

InChI

1S/C7H3Cl2N/c8-6-2-1-5(4-10)3-7(6)9/h1-3H

InChI key

KUWBYWUSERRVQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dichlorobenzonitrile (3,4-DCBN) is an aromatic nitrile that can be prepared from 3,4-dichlorobenzamide.[1]

Application

3,4-Dichlorobenzonitrilemay be used in the preparation of:
  • 3,4-difluorobenzonitrile[1]
  • 3-chloro-4-fluorobenzonitrile[1]
  • 5-chloromethyl-3-( 3,4-dichlorophenyl)isoxazole[2]
  • N-[3-(2-pyridyl)isoquinolin-1-yl]-3,4-dichlorobenzamidine[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and copper-dependent antimycoplasmal activity of 1-amino-3-(2-pyridyl) isoquinoline derivatives. 2. Amidines.
De Zwart MAH, et al.
Journal of Medicinal Chemistry, 32(2), 487-493 (1989)
3-Aryl-5-halomethylisoxazoles. A new class of anthelmintics.
Sen HG, et al.
Journal of Medicinal Chemistry, 9(3), 431-433 (1966)
Synthesis of 3, 4-difluorobenzonitrile and monofluorobenzonitriles by means of halogen-exchange fluorination.
Suzuki H and Kimura Y.
Journal of Fluorine Chemistry, 52(3), 341-351 (1991)
Wouter Huiting et al.
eLife, 11 (2022-02-25)
A loss of the checkpoint kinase ataxia telangiectasia mutated (ATM) leads to impairments in the DNA damage response, and in humans causes cerebellar neurodegeneration, and an increased risk of cancer. A loss of ATM is also associated with increased protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service