Key Documents
BR782891
BRAND® BRANDplates® for inserts
6-well with lid cellGrade plus
Synonym(s):
BRANDplates®, Cell culture, Insert System, Inserts, Plates, Tissue culture
Select a Size
10 EA
€130.00
€130.00
Check Cart for Availability
Select a Size
Change View
10 EA
€130.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material:
transparent wells
size:
6 well
€130.00
Check Cart for Availability
material
transparent wells
sterility
sterile
feature
lid
single packed
packaging
pack of 1 ea
pkg of Individually wrapped, with lid
manufacturer/tradename
BRAND 782891
size
6 well
suitability
suitable for (bacteriological assays)
General description
PS. cellGrade plus surface, sterile, hydrophilic with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible.Cover with condensation rings.Manufactured according to the requirements in ANSI/SLAS Standards 1 and 4.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Features and Benefits
  • Optimal cell growth thanks to cellGrade plus surface
  • Side well access for easier pipetting and removing cover slips
  • Perfect positioning of the BRANDplates® insert
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)