Skip to Content
Merck
All Photos(1)

Key Documents

BR781782

BRAND® microplate BRANDplates®, hydroGrade

size 96 wells, size 350 μL, flat bottom, transparent

Synonym(s):

BRANDplates®, Immunology, Microplates, hydroGrade

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€204.00

€204.00


Check Cart for Availability


Select a Size

Change View
100 EA
€204.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
colorless wells
flat bottom
polystyrene
size:
350 μL
96 wells

€204.00


Check Cart for Availability

material

colorless wells
flat bottom
polystyrene

sterility

non-sterile

feature

lid: no
skirt (U-bottom)

packaging

pack of 100 ea

manufacturer/tradename

BRAND 781782

size

350 μL
96 wells

working volume

350 μL

color

transparent

suitability

suitable for (bacteriological assays)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

For the immobilization of hydrophilic molecules.Strongly hydrophilic, with high affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids.An alternative to the immunoGrade surface when performing solid phase assays.Alternative for homogeneous assays with hydrophobic molecules, that remain in solution.PS, non-sterile.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service