BR781314
BRAND® PCR tubes
capacity 0.5 mL, blue tube, individual, attached flat cap
Synonym(s):
PCR, PCR Tubes, Tubes
Select a Size
€55.10
Select a Size
About This Item
€55.10
Recommended Products
material
blue cap
blue tube
conical bottom
flat cap
polypropylene cap
polypropylene tube
description
thin-walled
sterility
non-sterile
feature
graduations: no
individual, attached flat cap
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 781314
technique(s)
PCR: suitable
capacity
0.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Thin walls for rapid heat transfer
- Tightly closing covers protect samples against evaporation
- Colors available for easy identification of samples
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service