Skip to Content
Merck
All Photos(1)

Key Documents

BR704270

BRAND® thread adapter for remote dispensing system Dispensette®

stainless steel, O.D. 2 in.

Synonym(s):

Accessories, Bottle-top Dispenser, Dispenser, Dispensette®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€71.50

€71.50


Check Cart for Availability


Select a Size

Change View
1 EA
€71.50

About This Item

UNSPSC Code:
41121815
NACRES:
NB.01

€71.50


Check Cart for Availability

material

stainless steel

packaging

pack of 1 ea

manufacturer/tradename

BRAND 704270

O.D.

2 in.
50.8 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dispense accurate volumes directly from drums and bulk refills.The Dispensette® can be mounted on a wall, a ring stand or on lab furniture.A filter in the drum adapter minimizes risk of contaminating highpurity reagents when air is drawn in for pressure compensation after dispensing.A quick-release connector with integrated valves allows quick changing of the bulk container.The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette®.The max. delivery height is approximately 1.2 m.Operating Exclusions: Never use the remote dispensing system: 1. with recirculation valve. It has to be removed before use! 2. for pressurized vessels 3. for liquids attacking borosilicate glass, Al2O3-ceramic, PFA, ETFE, FEP or PTFE 4. for Peroxide (due to catalytic reaction) 5. for carbon disulfide (CS2), due to risk of explosion!

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service