Key Documents
BR4630330
BRAND® Dispensette® S Organic, Digital bottle-top dispenser
volume range 0.5-5 mL, without recirculation valve
Select a Size
1 EA
€405.00
€405.00
Check Cart for Availability
Select a Size
Change View
1 EA
€405.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€405.00
Check Cart for Availability
feature
incremental 0.02 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4630330
volume range
0.5-5 mL
fitting
(GL 28/S 28)
GL 32
GL 38
S 40
General description
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.