Key Documents
BR4600221
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 2 mL, with recirculation valve
Synonym(s):
Fixed-Volume Dispenser
Select a Size
1 EA
€404.00
€404.00
Check Cart for Availability
Select a Size
Change View
1 EA
€404.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€404.00
Check Cart for Availability
feature
DE-M marking
with recirculation valve
manufacturer/tradename
BRAND 4600221
volume
2 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.