Key Documents
BR144620
BRAND® BISTABIL® ground glass stopper, conical joint
borosilicate glass 3.3, joint: ST/NS 14/23, (hollow, hexagonal grip)
Select a Size
1 EA
€8.40
€8.40
Check Cart for Availability
Select a Size
Change View
1 EA
€8.40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€8.40
Check Cart for Availability
material
borosilicate glass 3.3
Joint
joint: ST/NS14/23
manufacturer/tradename
BRAND 144620
Related Categories
General description
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)