Skip to Content
Merck
All Photos(1)

Documents

BMSZ207A

Benchmark Sprint 8 Plus Clinical Centrifuge with fixed angle rotor

to hold, 8 x 15 mL, AC/DC input 115 V

Synonym(s):

Laboratory centrifuge, lab centrifuge, clinical centrifuge, blood centrifuge, urine centrifuge, 15ml centrifuge, Low speed centrifuge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701
NACRES:
NB.13

feature

refrigerated: no
to hold 8 x 15 mL

manufacturer/tradename

Benchmark Scientific Z207-A

parameter

6800 rpm speed

Looking for similar products? Visit Product Comparison Guide

General description

Unlike traditional clinical centrifuges the increased speed capability (4445xg) of the Sprint TM 8 PLUS allows for the separation of serum or plasma in as little as 23 minutes. Additional features include:

A large backlit ,liquid crystal display
High capacity rotor, 8 tubes up to 15ml (conical or round bottom)
Unique EZScroll touch pad
Timer up to 99hr99mins. / continuous
Storage of commonly used runtimes
Control of speed in both rpm and rcf
Adjustable acceleration and deceleration levels
Safety imbalance detection system
Momentary operation Electrical lid lock

Ideal for rapid production of serum and plasma

Application

Centrifugation of clinical samples including urine and blood. Ideal for production of PPP and PRP.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service