Skip to Content
Merck
All Photos(1)

Key Documents

BMSV10021SL

Sigma-Aldrich

Aspire Laboratory Aspirator Accessories

Optional single channel adapter, extra long (80 mm) stainless steel

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€42.90

€42.90


Check Cart for Availability


Select a Size

Change View
1 EA
€42.90

About This Item

UNSPSC Code:
41100000
NACRES:
NB.79

€42.90


Check Cart for Availability

manufacturer/tradename

Benchmark Scientific V1002-1SL

Looking for similar products? Visit Product Comparison Guide

General description

Accessories designed for use with Aspire laboratory aspirator.
The aspirator single channel 80mm long adapter is manufactured from stainless steel and enables aspiration from deep containers.

Legal Information

6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service