Key Documents
CLS4931
Costar® vacuum aspirator
8-channel adapter
Synonym(s):
Costar aspirator, Vacuum aspirator
Select a Size
1 EA
€299.00
€299.00
Check Cart for Availability
Select a Size
Change View
1 EA
€299.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.11
€299.00
Check Cart for Availability
packaging
pack of 1
manufacturer/tradename
Corning 4931
Related Categories
General description
This is an 8-channel adapter that fits on to the Costar® Aspirator (CLS4930) so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate.
Please note: Only the 8-channel adapter is supplied for product CLS4931, despite the image on this web page showing both the adapter and the Costar® Aspirator (CLS4930) together they are sold separately.
Legal Information
Costar is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)