BAH420530002
SP Bel-Art® Dry-Keeper Small, Stacking Gas-Purge Desiccator Cabinet
polystyrene, volume 0.1 ft3
All Photos(1)
About This Item
Recommended Products
material
polystyrene
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H42053-0002
volume
0.1 ft3
General description
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of lab bench space.
- Pull down door slides out of the way into a compartment beneath the cabinet
- Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
- Interior volume: 0.1 cu. ft
- Gas ports can be closed to use desiccator with standard desiccant cartridges
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP